Product Details

SNP ID
rs113820463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:108755936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGAGGAATTTTTCTAGAGGCAG[A/C]CACTCTTGTGTCTTACCAAGGTTAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00222 PubMed Links

Gene Details

Gene
LINC00222
Gene Name
long intergenic non-protein coding RNA 222
There are no transcripts associated with this gene.

View Full Product Details