Product Details

SNP ID
rs116503391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:118970580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAATATTAAGAAAATAATATAC[A/G]TAGCCAGAAATGACCCAATTCCCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM184A PubMed Links

Gene Details

Gene
FAM184A
Gene Name
family with sequence similarity 184 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100411.2 Intron NP_001093881.1
NM_001288576.1 Intron NP_001275505.1
NM_024581.5 Intron NP_078857.5

View Full Product Details