Product Details

SNP ID
rs116377893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:124963149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTGAACCCTCCCAGCACCCGCT[C/G]TTCCTTCCCCAGCCCCCGACTGTCC
Phenotype
MIM: 602532
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RNF217 PubMed Links

Gene Details

Gene
RNF217
Gene Name
ring finger protein 217
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286398.2 740 Missense Mutation TCT,TGT S202C NP_001273327.1
NM_152553.4 740 Intron NP_689766.1
XM_011535494.2 740 Missense Mutation TCT,TGT S202C XP_011533796.1
XM_011535495.2 740 Missense Mutation TCT,TGT S202C XP_011533797.1
XM_011535496.2 740 Missense Mutation TCT,TGT S202C XP_011533798.1
XM_011535499.2 740 Missense Mutation TCT,TGT S202C XP_011533801.1
XM_011535500.2 740 Intron XP_011533802.1
XM_017010317.1 740 Missense Mutation TCT,TGT S202C XP_016865806.1
Gene
RNF217-AS1
Gene Name
RNF217 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details