Product Details

SNP ID
rs117834537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132296831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAAAGTGAAATAGCAACAATGA[A/G]TATTTAAATAAAACAGAATGTAGTA
Phenotype
MIM: 609000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MOXD1 PubMed Links

Gene Details

Gene
MOXD1
Gene Name
monooxygenase DBH like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015529.3 2272 UTR 3 NP_056344.2
XM_017010714.1 2272 UTR 3 XP_016866203.1
XM_017010715.1 2272 UTR 3 XP_016866204.1

View Full Product Details