Product Details

SNP ID
rs116259921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:155230462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTAGCTCTCTCTGTTGTGTTTCC[A/G]TAGGACCCCCCACTTTCGGCACTGG
Phenotype
MIM: 607033 MIM: 604709
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TFB1M PubMed Links

Gene Details

Gene
TFB1M
Gene Name
transcription factor B1, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016020.3 Intron NP_057104.2
XM_005267006.4 Intron XP_005267063.1
XM_011535870.1 Intron XP_011534172.1
XM_011535871.1 Intron XP_011534173.1
XM_011535872.2 Intron XP_011534174.1
XM_011535873.2 Intron XP_011534175.1
XM_017010917.1 Intron XP_016866406.1
Gene
TIAM2
Gene Name
T-cell lymphoma invasion and metastasis 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010927.2 Intron NP_001010927.1
NM_012454.3 Intron NP_036586.2

View Full Product Details