Product Details
- SNP ID
-
rs117022509
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:31292875 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATTACAGGGATGAGCCACTGTGC[C/G]GGGCCAAAGCAGAATTTTAAATCAG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HEATR5A
PubMed Links
Gene Details
- Gene
- HEATR5A
- Gene Name
- HEAT repeat containing 5A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015473.3 |
6756 |
UTR 3 |
|
|
NP_056288.2 |
View Full Product Details