Product Details

SNP ID
rs115050412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:34712830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGACTGAGCTGGAGAAATGGAA[A/G]CTCCTTAAGATCCAGATGGCACTTG
Phenotype
MIM: 601443
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFL2 PubMed Links

Gene Details

Gene
CFL2
Gene Name
cofilin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243645.1 615 UTR 3 NP_001230574.1
NM_021914.7 615 UTR 3 NP_068733.1
NM_138638.4 615 UTR 3 NP_619579.1
XM_011536363.2 615 UTR 3 XP_011534665.1

View Full Product Details