Product Details

SNP ID
rs113421241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52553149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCGCCCGGCCCCCAGCCCCCTCA[G/T]GAGGGTGTCCGTGCCGGGTCCGGCG
Phenotype
MIM: 616179
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR137C PubMed Links

Gene Details

Gene
GPR137C
Gene Name
G protein-coupled receptor 137C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099652.1 2 Missense Mutation AGG,ATG R1M NP_001093122.1
Gene
TXNDC16
Gene Name
thioredoxin domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160047.1 2 Intron NP_001153519.1
NM_020784.2 2 Intron NP_065835.2
XM_017021505.1 2 Intron XP_016876994.1

View Full Product Details