Product Details

SNP ID
rs115935300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:54940751 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGGACTGCAATACTGTCACAGC[A/C]GTGTCTTCAGTTCTTCTCTACTGTG
Phenotype
MIM: 608126
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
WDHD1 PubMed Links

Gene Details

Gene
WDHD1
Gene Name
WD repeat and HMG-box DNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008396.2 4126 UTR 3 NP_001008397.1
NM_007086.3 4126 UTR 3 NP_009017.1
XM_006720012.2 4126 UTR 3 XP_006720075.1
XM_011536373.2 4126 UTR 3 XP_011534675.1
XM_017020940.1 4126 UTR 3 XP_016876429.1
XM_017020941.1 4126 UTR 3 XP_016876430.1

View Full Product Details