Product Details

SNP ID
rs117148720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:58202919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTAGTGTGATAAAAAGAGCTGGG[A/C]TGCCTAAGGTATTTAAAAAAAAATA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACTR10 PubMed Links

Gene Details

Gene
ACTR10
Gene Name
actin-related protein 10 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018477.2 201 Missense Mutation ATG,CTG M48L NP_060947.1
XM_011536960.1 201 Missense Mutation ATG,CTG M48L XP_011535262.1
XM_011536961.1 201 Missense Mutation ATG,CTG M48L XP_011535263.1
XM_011536962.2 201 UTR 5 XP_011535264.1
XM_011536963.2 201 UTR 5 XP_011535265.1

View Full Product Details