Product Details

SNP ID
rs115679402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:69383316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTATTTATTCCATTTCCACCTA[C/T]GCTGTTTAAAAAGAATTAATTTAAA
Phenotype
MIM: 601191 MIM: 615132
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERH PubMed Links

Gene Details

Gene
ERH
Gene Name
enhancer of rudimentary homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004450.2 Intron NP_004441.1
Gene
GALNT16
Gene Name
polypeptide N-acetylgalactosaminyltransferase 16
There are no transcripts associated with this gene.

View Full Product Details