Product Details

SNP ID
rs112860810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77181775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGGGGCTGGCTTCGCCCAGCC[A/G]GCGAGGAGGAGGACCCGCTGGCAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM63C PubMed Links

Gene Details

Gene
TMEM63C
Gene Name
transmembrane protein 63C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020431.3 17 UTR 5 NP_065164.2

View Full Product Details