Product Details

SNP ID
rs114497878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94462875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCCCATGAAGCTAGTTACCTGG[A/G]AGAATTTGTGGAAAAGGGCACTGAC
Phenotype
MIM: 615677
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINA9 PubMed Links

Gene Details

Gene
SERPINA9
Gene Name
serpin family A member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042518.1 1603 UTR 3 NP_001035983.1
NM_001284275.1 1603 UTR 3 NP_001271204.1
NM_001284276.1 1603 UTR 3 NP_001271205.1
NM_175739.3 1603 UTR 3 NP_783866.2
XM_011536714.2 1603 UTR 3 XP_011535016.1
XM_011536715.2 1603 UTR 3 XP_011535017.1
XM_011536716.2 1603 UTR 3 XP_011535018.1
XM_011536717.1 1603 UTR 3 XP_011535019.1

View Full Product Details