Product Details

SNP ID
rs113631765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:95701761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCCTTTTTGGCCTCTGACCTTT[C/T]AGTTGAAACAAAATTAGTTACCTAT
Phenotype
MIM: 186960 MIM: 603769
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCL1A PubMed Links

Gene Details

Gene
TCL1A
Gene Name
T-cell leukemia/lymphoma 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098725.1 Intron NP_001092195.1
NM_021966.2 Intron NP_068801.1
XM_017021676.1 Intron XP_016877165.1
XM_017021677.1 Intron XP_016877166.1
Gene
TCL1B
Gene Name
T-cell leukemia/lymphoma 1B
There are no transcripts associated with this gene.

View Full Product Details