Product Details

SNP ID
rs113174058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102365394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGAGGACTAGTTCTTACAGGGC[A/G]CATGTGGGGGAATAGGGAAGAGTGG
Phenotype
MIM: 613362 MIM: 615000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CINP PubMed Links

Gene Details

Gene
CINP
Gene Name
cyclin dependent kinase 2 interacting protein
There are no transcripts associated with this gene.

Gene
TECPR2
Gene Name
tectonin beta-propeller repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172631.2 Intron NP_001166102.1
NM_014844.4 Intron NP_055659.2

View Full Product Details