Product Details

SNP ID
rs116491072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:112651744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGGAAATTTGTACTATAAGTGG[A/G]AATTCTAGAGAAAGTGGCTAACTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP11AUN PubMed Links

Gene Details

Gene
ATP11AUN
Gene Name
ATP11A upstream neighbor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207440.2 Intron NP_997323.1

View Full Product Details