Product Details

SNP ID
rs115762779
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:147392743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCATCATGTCACTTCCACTCAC[C/T]TGGGGAGATTCTAGGACTGAGACAC
Phenotype
MIM: 601168
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DPYSL3 PubMed Links

Gene Details

Gene
DPYSL3
Gene Name
dihydropyrimidinase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197294.1 3545 UTR 3 NP_001184223.1
NM_001387.2 3545 UTR 3 NP_001378.1
XM_011537574.1 3545 UTR 3 XP_011535876.1
Gene
STK32A
Gene Name
serine/threonine kinase 32A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112724.1 3545 Intron NP_001106195.1
NM_001287740.1 3545 Intron NP_001274669.1
NM_145001.3 3545 Intron NP_659438.1
XM_011537576.1 3545 Intron XP_011535878.1
XM_011537577.2 3545 Intron XP_011535879.1
XM_011537579.2 3545 Intron XP_011535881.1
XM_017009213.1 3545 Intron XP_016864702.1
XM_017009214.1 3545 Intron XP_016864703.1

View Full Product Details