Product Details

SNP ID
rs115782087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48905046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGAAGTCAAGACCAGAGAACTTC[A/T]TAAGGACATTGAGATCCTCAGCCAA
Phenotype
MIM: 611088
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC65 PubMed Links

Gene Details

Gene
CCDC65
Gene Name
coiled-coil domain containing 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286957.1 460 UTR 5 NP_001273886.1
NM_033124.4 460 Missense Mutation CAT,CTT H78L NP_149115.2

View Full Product Details