Product Details
- SNP ID
-
rs112889908
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52360018 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGACCACACGCTCTTGCATTGCAGC[A/G]GCATTTATTGAAACACAGATCGAGA
- Phenotype
-
MIM: 602767
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT85
PubMed Links
Gene Details
- Gene
- KRT85
- Gene Name
- keratin 85
View Full Product Details