Product Details

SNP ID
rs116694250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54609380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTCACCTCCATGAGCTGAAAC[C/T]TTTTACCTGCTAGCACTTGGGTGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLYCAM1 PubMed Links

Gene Details

Gene
GLYCAM1
Gene Name
glycosylation dependent cell adhesion molecule 1 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details