Product Details

SNP ID
rs115311663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56086524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTAACCCTGTCACTTCTTTCCCT[A/T]CCTCAGAGATTCTGTCAGGGGGTGT
Phenotype
MIM: 190151
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ERBB3 PubMed Links

Gene Details

Gene
ERBB3
Gene Name
erb-b2 receptor tyrosine kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005915.1 Intron NP_001005915.1
NM_001982.3 Intron NP_001973.2

View Full Product Details