Product Details

SNP ID
rs114910664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56599688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTTTGAGTGTGGGAAAGAAAG[A/C]GCAGAAGCCTTACCTGAGCCAAACA
Phenotype
MIM: 605682 MIM: 602387
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BAZ2A PubMed Links

Gene Details

Gene
BAZ2A
Gene Name
bromodomain adjacent to zinc finger domain 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300905.1 Intron NP_001287834.1
NM_013449.3 Intron NP_038477.2
XM_011537797.2 Intron XP_011536099.1
XM_011537798.2 Intron XP_011536100.1
XM_011537799.2 Intron XP_011536101.1
XM_011537800.2 Intron XP_011536102.1
XM_011537801.2 Intron XP_011536103.1
XM_011537803.2 Intron XP_011536105.1
XM_011537804.2 Intron XP_011536106.1
XM_011537805.2 Intron XP_011536107.1
XM_011537806.2 Intron XP_011536108.1
XM_011537807.2 Intron XP_011536109.1
XM_011537809.2 Intron XP_011536111.1
XM_017018740.1 Intron XP_016874229.1
XM_017018741.1 Intron XP_016874230.1
XM_017018742.1 Intron XP_016874231.1
Gene
RBMS2
Gene Name
RNA binding motif single stranded interacting protein 2
There are no transcripts associated with this gene.

View Full Product Details