Product Details

SNP ID
rs111985538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56670298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCAGAGAAACGATCAAAATTAG[A/G]CATGTCTTCATCTGAATCATCTTCC
Phenotype
MIM: 607061
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTGES3 PubMed Links

Gene Details

Gene
PTGES3
Gene Name
prostaglandin E synthase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282601.1 761 Missense Mutation CCT,TCT P118S NP_001269530.1
NM_001282602.1 761 Intron NP_001269531.1
NM_001282603.1 761 Missense Mutation CCT,TCT P85S NP_001269532.1
NM_001282604.1 761 Missense Mutation CCT,TCT P122S NP_001269533.1
NM_001282605.1 761 Intron NP_001269534.1
NM_006601.6 761 Missense Mutation CCT,TCT P118S NP_006592.3
XM_005268576.4 761 Missense Mutation CCT,TCT P122S XP_005268633.1
XM_006719199.2 761 Intron XP_006719262.1
XM_011537773.2 761 Intron XP_011536075.1
XM_011537774.2 761 Missense Mutation CCT,TCT P122S XP_011536076.1
XM_017018716.1 761 Missense Mutation CCT,TCT P122S XP_016874205.1

View Full Product Details