Product Details
- SNP ID
-
rs115691862
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:65051385 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTGGATTCAGGTGGATCCCGCCG[C/T]TCCTCGGCCTTTTTAAGTGAAGGCG
- Phenotype
-
MIM: 605186
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WIF1
PubMed Links
Gene Details
- Gene
- WIF1
- Gene Name
- WNT inhibitory factor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007191.4 |
1479 |
Silent Mutation |
GAA,GAG |
E368E |
NP_009122.2 |
View Full Product Details