Product Details

SNP ID
rs115205809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:65051632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAATGAAGAGATCTCTGTTAAG[A/G]GCAACCCATTTTACTAACTCCATGA
Phenotype
MIM: 605186
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WIF1 PubMed Links

Gene Details

Gene
WIF1
Gene Name
WNT inhibitory factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007191.4 Intron NP_009122.2

View Full Product Details