Product Details

SNP ID
rs117820493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:45176771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGAAACTTAACCCTGGTAAGCT[C/T]ATTCAAACTTCCCAAGTCTCCCTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLEKHA8P1 PubMed Links

Gene Details

Gene
PLEKHA8P1
Gene Name
pleckstrin homology domain containing A8 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details