Product Details

SNP ID
rs114814977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:197674544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCTCCTCGGGGCACAGTCTGAC[A/C]CCAGTCCAGGACAGGGAGAGGGAAA
Phenotype
MIM: 613516
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIR922 PubMed Links

Gene Details

Gene
MIR922
Gene Name
microRNA 922
There are no transcripts associated with this gene.

Gene
RUBCN
Gene Name
RUN and cysteine rich domain containing beclin 1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145642.3 3716 UTR 3 NP_001139114.1
NM_014687.2 3716 UTR 3 NP_055502.1
XM_005269374.3 3716 UTR 3 XP_005269431.1
XM_006713827.3 3716 UTR 3 XP_006713890.1
XM_006713828.3 3716 UTR 3 XP_006713891.1
XM_006713829.3 3716 UTR 3 XP_006713892.1
XM_006713830.3 3716 UTR 3 XP_006713893.1
XM_006713831.3 3716 UTR 3 XP_006713894.1
XM_017007543.1 3716 UTR 3 XP_016863032.1
XM_017007544.1 3716 UTR 3 XP_016863033.1
XM_017007545.1 3716 UTR 3 XP_016863034.1

View Full Product Details