Product Details

SNP ID
rs117590923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:108224307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTAGAAAAGAATGGCATGCCAA[A/G]GGCAGAGTTATTTCTAGGAAGATGG
Phenotype
MIM: 607585 MIM: 601448
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATM PubMed Links

Gene Details

Gene
ATM
Gene Name
ATM serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000051.3 Intron NP_000042.3
XM_005271561.4 Intron XP_005271618.2
XM_005271562.4 Intron XP_005271619.2
XM_006718843.3 Intron XP_006718906.1
XM_006718845.1 Intron XP_006718908.1
XM_011542840.2 Intron XP_011541142.1
XM_011542842.2 Intron XP_011541144.1
XM_011542843.2 Intron XP_011541145.1
XM_011542844.2 Intron XP_011541146.1
XM_011542845.1 Intron XP_011541147.1
XM_017017789.1 Intron XP_016873278.1
XM_017017790.1 Intron XP_016873279.1
XM_017017791.1 Intron XP_016873280.1
XM_017017792.1 Intron XP_016873281.1
Gene
NPAT
Gene Name
nuclear protein, coactivator of histone transcription
There are no transcripts associated with this gene.

View Full Product Details