Product Details

SNP ID
rs117941637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114059818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGTTCCCCAGCAACACCCCTCC[C/T]CGACACAGGCACACACCCCCCGACA
Phenotype
MIM: 176797
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZBTB16 PubMed Links

Gene Details

Gene
ZBTB16
Gene Name
zinc finger and BTB domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018011.1 242 Intron NP_001018011.1
NM_006006.4 242 UTR 5 NP_005997.2
XM_005271658.4 242 UTR 5 XP_005271715.1
XM_017018257.1 242 Intron XP_016873746.1
XM_017018258.1 242 Intron XP_016873747.1
XM_017018259.1 242 Intron XP_016873748.1

View Full Product Details