Product Details

SNP ID
rs116572317
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114296389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACAGCCAGAACATTTGTGGTCTA[C/T]TTCTCTGTTAGTGTTTAACCAACCA
Phenotype
MIM: 600008
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NNMT PubMed Links

Gene Details

Gene
NNMT
Gene Name
nicotinamide N-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006169.2 577 UTR 5 NP_006160.1

View Full Product Details