Product Details

SNP ID
rs118176702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:122838802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGTTCAATGGCTAAGGAGTATAG[A/C]AAGGATCATTATAGTGTGTGTCTCT
Phenotype
MIM: 612597
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CRTAM PubMed Links

Gene Details

Gene
CRTAM
Gene Name
cytotoxic and regulatory T-cell molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304782.1 Intron NP_001291711.1
NM_019604.3 Intron NP_062550.2
XM_011542900.2 Intron XP_011541202.1

View Full Product Details