Product Details

SNP ID
rs111755132
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124570154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAATCACTCTACAGTGACAGAGT[C/T]CATTCTCAAGGGTTTAACGAAGAGA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR8A1 PubMed Links

Gene Details

Gene
OR8A1
Gene Name
olfactory receptor family 8 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005194.1 86 Missense Mutation TCC,TTC S29F NP_001005194.1

View Full Product Details