Product Details

SNP ID
rs117126817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:100993027 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGAGACCATTTTCATTTACTGCC[A/T]TCATAAATATTCTCCACCATTCAAG
Phenotype
MIM: 615936
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP42 PubMed Links

Gene Details

Gene
ARHGAP42
Gene Name
Rho GTPase activating protein 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152432.2 948 Intron NP_689645.2
XM_011542615.2 948 UTR 3 XP_011540917.1
XM_011542616.2 948 UTR 3 XP_011540918.1
XM_011542617.2 948 UTR 3 XP_011540919.1
XM_017017237.1 948 UTR 3 XP_016872726.1
XM_017017238.1 948 UTR 3 XP_016872727.1
Gene
TMEM133
Gene Name
transmembrane protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032021.2 948 UTR 3 NP_114410.1

View Full Product Details