Product Details

SNP ID
rs114775338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:109338064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACAACATTCTAGGACCAATGGA[A/C]GATTATAGAGCTACGTTATCTGATA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC285638 PubMed Links

Gene Details

Gene
LOC285638
Gene Name
uncharacterized LOC285638
There are no transcripts associated with this gene.

Gene
PJA2
Gene Name
praja ring finger ubiquitin ligase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014819.4 Intron NP_055634.3
XM_017010099.1 Intron XP_016865588.1

View Full Product Details