Product Details

SNP ID
rs114067422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:111094205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATAGGTGGTGGTAGTGATGGTAG[C/T]AGCTATCATGTATTGAGCAGTTACT
Phenotype
MIM: 609669
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR36 PubMed Links

Gene Details

Gene
WDR36
Gene Name
WD repeat domain 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139281.2 Intron NP_644810.1

View Full Product Details