Product Details

SNP ID
rs116481172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:135935637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACGTACAAGATGACCATATATAC[C/T]TGGACGTATCTGGACTTCAAATTTG
Phenotype
MIM: 609087
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXL21 PubMed Links

Gene Details

Gene
FBXL21
Gene Name
F-box and leucine rich repeat protein 21 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012159.4 394 UTR 5 NP_036291.2

View Full Product Details