Product Details

SNP ID
rs115362122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138364927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACTTCACATTTAATTTATTCAGG[A/G]TGTTAGAAAGTAGAAAACTTTGGGG
Phenotype
MIM: 609373
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDM3B PubMed Links

Gene Details

Gene
KDM3B
Gene Name
lysine demethylase 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016604.3 Intron NP_057688.2
XM_005272018.4 Intron XP_005272075.1
XM_011543488.2 Intron XP_011541790.1
XM_011543489.2 Intron XP_011541791.1
XM_017009584.1 Intron XP_016865073.1

View Full Product Details