Product Details

SNP ID
rs117166939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7604343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCCTATTTAGGGACCTCAGGCT[A/G]TCTCTATGATGTGACCTCACCATCT
Phenotype
MIM: 612685
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMSAP3 PubMed Links

Gene Details

Gene
CAMSAP3
Gene Name
calmodulin regulated spectrin associated protein family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080429.2 Intron NP_001073898.1
NM_020902.1 Intron NP_065953.1
XM_005272491.3 Intron XP_005272548.1
XM_011528154.2 Intron XP_011526456.1
XM_017027036.1 Intron XP_016882525.1

View Full Product Details