Product Details

SNP ID
rs116398041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7961182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGGGGGCGAGGAAAGAAGCTGA[A/G]GTGCTACAAGCCCGTCATCATTTTC
Phenotype
MIM: 603466
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELAVL1 PubMed Links

Gene Details

Gene
ELAVL1
Gene Name
ELAV like RNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001419.2 3449 UTR 3 NP_001410.2

View Full Product Details