Product Details
- SNP ID
-
rs111966921
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:41183581 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCTCACTTCACTGAACAGCAGAG[A/G]CAACCGTTTCTAAGTTCCAGCCACT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CBWD6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs62557779] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CBWD6
- Gene Name
- COBW domain containing 6
View Full Product Details