Product Details

SNP ID
rs104894168
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73250935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACTTCACGTGCCCGTTTCCTTC[G/A]CAGTCTCTCAGCATTTGTGATCATC
Phenotype
MIM: 611206 MIM: 609204
Polymorphism
G/A, Transition Substitution
Allele Nomenclature
Literature Links
CFAP70 PubMed Links

Gene Details

Gene
CFAP70
Gene Name
cilia and flagella associated protein 70
There are no transcripts associated with this gene.

Gene
DNAJC9
Gene Name
DnaJ heat shock protein family (Hsp40) member C9
There are no transcripts associated with this gene.

Gene
DNAJC9-AS1
Gene Name
DNAJC9 antisense RNA 1
There are no transcripts associated with this gene.

Gene
FAM149B1
Gene Name
family with sequence similarity 149 member B1
There are no transcripts associated with this gene.

Gene
MRPS16
Gene Name
mitochondrial ribosomal protein S16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016065.3 Intron NP_057149.1
XM_017016305.1 Intron XP_016871794.1

View Full Product Details