Product Details

SNP ID
rs111794597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:132333292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACAAAGTGAGAGCAGAGAATTT[C/T]AATCTGAACCTCTTTGTATTCCTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC27 PubMed Links

Gene Details

Gene
LRRC27
Gene Name
leucine rich repeat containing 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143757.1 Intron NP_001137229.1
NM_001143758.1 Intron NP_001137230.1
NM_001143759.1 Intron NP_001137231.1
NM_001309474.1 Intron NP_001296403.1
NM_030626.2 Intron NP_085129.1
XM_006717983.2 Intron XP_006718046.1
XM_006717984.2 Intron XP_006718047.1
XM_011540208.1 Intron XP_011538510.1
XM_011540209.1 Intron XP_011538511.1
XM_011540210.2 Intron XP_011538512.1
XM_017016690.1 Intron XP_016872179.1
XM_017016691.1 Intron XP_016872180.1
XM_017016692.1 Intron XP_016872181.1
XM_017016693.1 Intron XP_016872182.1
XM_017016694.1 Intron XP_016872183.1
Gene
STK32C
Gene Name
serine/threonine kinase 32C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318878.1 Intron NP_001305807.1
NM_001318879.1 Intron NP_001305808.1
NM_001318880.1 Intron NP_001305809.1
NM_001318881.1 Intron NP_001305810.1
NM_001318882.1 Intron NP_001305811.1
NM_001318883.1 Intron NP_001305812.1
NM_173575.3 Intron NP_775846.2
XM_011539688.1 Intron XP_011537990.1
XM_011539690.2 Intron XP_011537992.1
XM_011539693.2 Intron XP_011537995.1
XM_011539694.1 Intron XP_011537996.1
XM_011539695.1 Intron XP_011537997.1
XM_011539696.1 Intron XP_011537998.1
XM_017016100.1 Intron XP_016871589.1
XM_017016101.1 Intron XP_016871590.1

View Full Product Details