Product Details

SNP ID
rs111800178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:86960116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAGAACATCGCGGTCACCTCCGG[A/G]GTGGTGCGCAAGGTGAGCCCCGGCC
Phenotype
MIM: 608925 MIM: 602998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADIRF PubMed Links

Gene Details

Gene
ADIRF
Gene Name
adipogenesis regulatory factor
There are no transcripts associated with this gene.

Gene
MMRN2
Gene Name
multimerin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024756.2 446 Intron NP_079032.2
XM_005270153.1 446 Intron XP_005270210.1
XM_006717970.3 446 Intron XP_006718033.1
Gene
SNCG
Gene Name
synuclein gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003087.2 446 Silent Mutation GGA,GGG G93G NP_003078.2
XM_011540105.2 446 Missense Mutation AGT,GGT S111G XP_011538407.1

View Full Product Details