Product Details

SNP ID
rs112044835
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95313070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAATTCAGGAAGTCTCCTTCTAC[C/T]CAGAAAGCTCACCCCCTCCAGTTCC
Phenotype
MIM: 605264
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SORBS1 PubMed Links

Gene Details

Gene
SORBS1
Gene Name
sorbin and SH3 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034954.1 5606 UTR 3 NP_001030126.1
NM_001034955.1 5606 UTR 3 NP_001030127.1
NM_001034956.1 5606 UTR 3 NP_001030128.1
NM_001034957.1 5606 UTR 3 NP_001030129.1
NM_001290294.1 5606 UTR 3 NP_001277223.1
NM_001290295.1 5606 UTR 3 NP_001277224.1
NM_001290296.1 5606 UTR 3 NP_001277225.1
NM_001290297.1 5606 UTR 3 NP_001277226.1
NM_001290298.1 5606 UTR 3 NP_001277227.1
NM_006434.2 5606 UTR 3 NP_006425.2
NM_015385.3 5606 UTR 3 NP_056200.1
NM_024991.2 5606 UTR 3 NP_079267.1
XM_006717589.1 5606 UTR 3 XP_006717652.1
XM_006717593.1 5606 UTR 3 XP_006717656.1
XM_011539140.1 5606 UTR 3 XP_011537442.1
XM_011539150.1 5606 UTR 3 XP_011537452.1
XM_011539155.1 5606 UTR 3 XP_011537457.1
XM_011539167.1 5606 UTR 3 XP_011537469.1
XM_017015500.1 5606 UTR 3 XP_016870989.1
XM_017015501.1 5606 UTR 3 XP_016870990.1
XM_017015502.1 5606 UTR 3 XP_016870991.1
XM_017015503.1 5606 UTR 3 XP_016870992.1
XM_017015504.1 5606 UTR 3 XP_016870993.1
XM_017015505.1 5606 UTR 3 XP_016870994.1
XM_017015506.1 5606 UTR 3 XP_016870995.1
XM_017015507.1 5606 UTR 3 XP_016870996.1
XM_017015508.1 5606 UTR 3 XP_016870997.1
XM_017015509.1 5606 UTR 3 XP_016870998.1
XM_017015510.1 5606 UTR 3 XP_016870999.1
XM_017015511.1 5606 UTR 3 XP_016871000.1
XM_017015512.1 5606 UTR 3 XP_016871001.1
XM_017015513.1 5606 UTR 3 XP_016871002.1
XM_017015514.1 5606 UTR 3 XP_016871003.1
XM_017015515.1 5606 UTR 3 XP_016871004.1
XM_017015516.1 5606 UTR 3 XP_016871005.1
XM_017015517.1 5606 UTR 3 XP_016871006.1
XM_017015518.1 5606 UTR 3 XP_016871007.1
XM_017015519.1 5606 UTR 3 XP_016871008.1
XM_017015520.1 5606 UTR 3 XP_016871009.1
XM_017015521.1 5606 UTR 3 XP_016871010.1
XM_017015522.1 5606 UTR 3 XP_016871011.1
XM_017015523.1 5606 UTR 3 XP_016871012.1
XM_017015524.1 5606 UTR 3 XP_016871013.1
XM_017015525.1 5606 UTR 3 XP_016871014.1
XM_017015526.1 5606 UTR 3 XP_016871015.1
XM_017015527.1 5606 UTR 3 XP_016871016.1
XM_017015528.1 5606 UTR 3 XP_016871017.1
XM_017015529.1 5606 UTR 3 XP_016871018.1
XM_017015530.1 5606 UTR 3 XP_016871019.1
XM_017015531.1 5606 UTR 3 XP_016871020.1
XM_017015532.1 5606 UTR 3 XP_016871021.1
XM_017015533.1 5606 UTR 3 XP_016871022.1
XM_017015534.1 5606 UTR 3 XP_016871023.1
XM_017015535.1 5606 UTR 3 XP_016871024.1
XM_017015536.1 5606 UTR 3 XP_016871025.1
XM_017015537.1 5606 UTR 3 XP_016871026.1
XM_017015538.1 5606 UTR 3 XP_016871027.1
XM_017015539.1 5606 UTR 3 XP_016871028.1
XM_017015540.1 5606 UTR 3 XP_016871029.1

View Full Product Details