Product Details
- SNP ID
-
rs113153496
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:277469 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACCACTCCGACGATCAGGTAGTG[C/T]TCCTCCAGGACCACGTTGGTCACCA
- Phenotype
-
MIM: 611380
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DIP2C
PubMed Links
Gene Details
- Gene
- DIP2C
- Gene Name
- disco interacting protein 2 homolog C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014974.2 |
2908 |
Silent Mutation |
GAA,GAG |
E1509E |
NP_055789.1 |
XM_005252426.3 |
2908 |
Silent Mutation |
GAA,GAG |
E1565E |
XP_005252483.1 |
XM_005252427.3 |
2908 |
Silent Mutation |
GAA,GAG |
E1519E |
XP_005252484.1 |
XM_005252428.4 |
2908 |
Silent Mutation |
GAA,GAG |
E1575E |
XP_005252485.1 |
XM_005252430.2 |
2908 |
Silent Mutation |
GAA,GAG |
E946E |
XP_005252487.2 |
XM_011519428.2 |
2908 |
Silent Mutation |
GAA,GAG |
E1579E |
XP_011517730.1 |
XM_011519429.2 |
2908 |
Silent Mutation |
GAA,GAG |
E1569E |
XP_011517731.1 |
XM_011519430.2 |
2908 |
Silent Mutation |
GAA,GAG |
E1523E |
XP_011517732.1 |
XM_011519431.2 |
2908 |
Silent Mutation |
GAA,GAG |
E1513E |
XP_011517733.1 |
XM_011519432.1 |
2908 |
Silent Mutation |
GAA,GAG |
E1412E |
XP_011517734.1 |
XM_011519433.2 |
2908 |
Silent Mutation |
GAA,GAG |
E840E |
XP_011517735.1 |
XM_017015977.1 |
2908 |
Silent Mutation |
GAA,GAG |
E956E |
XP_016871466.1 |
View Full Product Details