Product Details

SNP ID
rs113773165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:62805508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACGGCCGCGGGCCTTGCCGCCCG[A/T]GCAGCAGTTCGAGCCGCCGCTGCAG
Phenotype
MIM: 611392 MIM: 129010
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ADO PubMed Links

Gene Details

Gene
ADO
Gene Name
2-aminoethanethiol dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032804.5 753 Missense Mutation GAG,GTG E150V NP_116193.2
Gene
EGR2
Gene Name
early growth response 2
There are no transcripts associated with this gene.

View Full Product Details