Product Details

SNP ID
rs113995300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:37950724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGATGTTGTCACTATCACTACTC[A/T]TTTATGGGATGGGAACTGAGGCACA
Phenotype
MIM: 194528
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF25 PubMed Links

Gene Details

Gene
ZNF25
Gene Name
zinc finger protein 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005252384.2 3117 UTR 3 XP_005252441.1
XM_005252385.2 3117 UTR 3 XP_005252442.1
XM_005252386.2 3117 UTR 3 XP_005252443.1
XM_011519352.2 3117 UTR 3 XP_011517654.1
XM_011519353.2 3117 UTR 3 XP_011517655.1
XM_011519354.2 3117 UTR 3 XP_011517656.1
XM_011519355.2 3117 UTR 3 XP_011517657.1
XM_011519356.2 3117 UTR 3 XP_011517658.1
XM_017015837.1 3117 UTR 3 XP_016871326.1
XM_017015838.1 3117 UTR 3 XP_016871327.1

View Full Product Details