Product Details

SNP ID
rs114220245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95910712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTAATCAATTTCCCTGCCTCC[A/G]TAGAATATCTATATTTAATCATTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf131 PubMed Links

Gene Details

Gene
C10orf131
Gene Name
chromosome 10 open reading frame 131
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130446.2 Intron NP_001123918.2
Gene
ENTPD1-AS1
Gene Name
ENTPD1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details