Product Details

SNP ID
rs114676717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:71713666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGGAGGAAGTAGAGGGTTCTCT[C/T]GATCAGGGCCTCAGCTTGTGAGGAC
Phenotype
MIM: 605516
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf105 PubMed Links

Gene Details

Gene
C10orf105
Gene Name
chromosome 10 open reading frame 105
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164375.2 2872 UTR 3 NP_001157847.1
NM_001168390.1 2872 UTR 3 NP_001161862.1
XM_011539808.2 2872 UTR 3 XP_011538110.1
XM_011539809.2 2872 UTR 3 XP_011538111.1
Gene
CDH23
Gene Name
cadherin-related 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171930.1 2872 Intron NP_001165401.1
NM_001171931.1 2872 Intron NP_001165402.1
NM_001171932.1 2872 Intron NP_001165403.1
NM_001171933.1 2872 Intron NP_001165404.1
NM_001171934.1 2872 Intron NP_001165405.1
NM_001171935.1 2872 Intron NP_001165406.1
NM_001171936.1 2872 Intron NP_001165407.1
NM_022124.5 2872 Intron NP_071407.4
NM_052836.3 2872 Intron NP_443068.1
XM_006717940.3 2872 Intron XP_006718003.1
XM_006717942.3 2872 Intron XP_006718005.1
XM_011540039.2 2872 Intron XP_011538341.1
XM_011540042.2 2872 Intron XP_011538344.1
XM_011540043.2 2872 Intron XP_011538345.1
XM_011540044.2 2872 Intron XP_011538346.1
XM_011540045.2 2872 Intron XP_011538347.1
XM_011540046.2 2872 Intron XP_011538348.1
XM_011540047.2 2872 Intron XP_011538349.1
XM_011540048.2 2872 Intron XP_011538350.1
XM_011540049.2 2872 Intron XP_011538351.1
XM_011540051.2 2872 Intron XP_011538353.1
XM_011540052.2 2872 Intron XP_011538354.1
XM_017016499.1 2872 Intron XP_016871988.1
XM_017016500.1 2872 Intron XP_016871989.1
XM_017016501.1 2872 Intron XP_016871990.1
XM_017016502.1 2872 Intron XP_016871991.1
XM_017016503.1 2872 Intron XP_016871992.1
XM_017016504.1 2872 Intron XP_016871993.1
XM_017016505.1 2872 Intron XP_016871994.1
XM_017016506.1 2872 Intron XP_016871995.1
XM_017016507.1 2872 Intron XP_016871996.1
XM_017016508.1 2872 Intron XP_016871997.1

View Full Product Details