Product Details

SNP ID
rs115222937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:63136512 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAAATTCATTGTATAAGGACTTA[C/T]CACAGTACATGGATTTGTAATCATG
Phenotype
MIM: 616477
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NRBF2 PubMed Links

Gene Details

Gene
NRBF2
Gene Name
nuclear receptor binding factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282405.1 Intron NP_001269334.1
NM_030759.4 Intron NP_110386.2
XM_006717810.3 Intron XP_006717873.1

View Full Product Details